Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIDA
Entrez Gene ID
101106862
Gene Name
axin interactor, dorsalization associated
Gene Aliases
-
Chromosome Location
Chr.12: 28546250 - 28595051 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013613.4 XP_004013662.3 1 - 2 276 66

Gene Symbol
LOC114112240
Entrez Gene ID
114112240
Gene Name
axin interactor, dorsalization-associated protein-like
Gene Aliases
-
Chromosome Location
Chr.Unknown: 12157 - 13584 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963759.1 XP_027819560.1 1 - 1 248 66

View Full Product Details