Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC9
Entrez Gene ID
101117902
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
Gene Aliases
-
Chromosome Location
Chr.25: 30343315 - 30348882 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962208.1 XP_027818009.1 3 - 4 1099 82

View Full Product Details