Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC17
Entrez Gene ID
100302359
Gene Name
thioredoxin domain containing 17
Gene Aliases
-
Chromosome Location
Chr.11: 38120361 - 38122812 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163057.1 NP_001156529.1 2 - 3 263 93
GenBank mRNA FJ969407.1 - 2 - 3 263 93

View Full Product Details