Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NODAL
Entrez Gene ID
101110384
Gene Name
nodal growth differentiation factor
Gene Aliases
-
Chromosome Location
Chr.25: 27552430 - 27560127 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021434.4 XP_004021483.4 1 - 2 197 64

View Full Product Details