Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYDE2
Entrez Gene ID
101112609
Gene Name
synapse defective Rho GTPase homolog 2
Gene Aliases
-
Chromosome Location
Chr.1: 66766270 - 66809425 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002146.4 XP_004002195.3 4 - 5 2638 93

View Full Product Details