Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KBTBD8
Entrez Gene ID
101118139
Gene Name
kelch repeat and BTB domain containing 8
Gene Aliases
-
Chromosome Location
Chr.19: 36188397 - 36200681 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018341.4 XP_004018390.1 3 - 4 1409 80

View Full Product Details