Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC16A2
Entrez Gene ID
101104210
Gene Name
solute carrier family 16 member 2
Gene Aliases
-
Chromosome Location
Chr.X: 75478582 - 75614479 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962830.1 XP_027818631.1 1 - 2 1169 102

View Full Product Details