Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL10
Entrez Gene ID
101118201
Gene Name
mitochondrial ribosomal protein L10
Gene Aliases
-
Chromosome Location
Chr.11: 24692420 - 24698454 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012816.3 XP_004012865.1 3 - 4 439 84

View Full Product Details