Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAPRE3
Entrez Gene ID
101112123
Gene Name
microtubule associated protein RP/EB family member 3
Gene Aliases
-
Chromosome Location
Chr.3: 35591960 - 35646934 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015094261.2 XP_014949747.1 3 - 4 464 138
XM_015094262.2 XP_014949748.1 2 - 3 412 138
XM_027966593.1 XP_027822394.1 3 - 4 443 138

View Full Product Details