Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA14
Entrez Gene ID
443315
Gene Name
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 14
Gene Aliases
-
Chromosome Location
Chr.18: 59513276 - 59533662 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009304.1 NP_001009304.1 3 - 4 948 80
XM_027956983.1 XP_027812784.1 3 - 4 937 80
GenBank mRNA AY679518.1 - 2 - 3 900 80
M21027.1 - 3 - 4 948 80

View Full Product Details