Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VIP
Entrez Gene ID
100145884
Gene Name
vasoactive intestinal peptide
Gene Aliases
-
Chromosome Location
Chr.8: 83836510 - 83845502 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126368.1 NP_001119840.1 2 - 3 228 107
XM_012182368.3 XP_012037758.1 3 - 4 475 107
GenBank mRNA EU016225.1 - 2 - 3 228 107

View Full Product Details