Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RILPL2
Entrez Gene ID
100302076
Gene Name
Rab interacting lysosomal protein like 2
Gene Aliases
-
Chromosome Location
Chr.17: 61165455 - 61194324 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161873.1 NP_001155345.1 3 - 4 630 98
GenBank mRNA FJ943981.1 - 3 - 4 630 98

View Full Product Details