Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCGR3A
Entrez Gene ID
100192433
Gene Name
Fc fragment of IgG, low affinity IIIa, receptor (CD16a)
Gene Aliases
-
Chromosome Location
Chr.1: 120125218 - 120136004 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139454.1 NP_001132926.1 2 - 3 77 76
XM_012188046.2 XP_012043436.2 2 - 3 76
GenBank mRNA EU589390.1 - 2 - 3 77 76

View Full Product Details