Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB21
Entrez Gene ID
100302077
Gene Name
RAB21, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.3: 114005544 - 114034063 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161874.1 NP_001155346.1 6 - 7 519 89
GenBank mRNA FJ943982.1 - 6 - 7 519 89

View Full Product Details