Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL5C
Entrez Gene ID
100302314
Gene Name
ADP ribosylation factor like GTPase 5C
Gene Aliases
-
Chromosome Location
Chr.11: 23513595 - 23521965 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162556.1 NP_001156028.1 1 - 2 49 61
XM_015098289.2 XP_014953775.1 2 - 3 1897 61
XM_027973939.1 XP_027829740.1 2 - 3 1897 61
XM_027973940.1 XP_027829741.1 2 - 3 1884 61
GenBank mRNA FJ969401.1 - 1 - 2 49 61

View Full Product Details