Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL5C
Entrez Gene ID
100302314
Gene Name
ADP ribosylation factor like GTPase 5C
Gene Aliases
-
Chromosome Location
Chr.11: 23513595 - 23521965 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162556.1 NP_001156028.1 2 - 3 111 73
XM_027973939.1 XP_027829740.1 3 - 4 1959 73
XM_027973940.1 XP_027829741.1 3 - 4 1946 73
GenBank mRNA FJ969401.1 - 2 - 3 111 73

View Full Product Details