Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LALBA
Entrez Gene ID
443386
Gene Name
lactalbumin alpha
Gene Aliases
-
Chromosome Location
Chr.3: 147491761 - 147493791 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009797.1 NP_001009797.1 1 - 2 158 76
GenBank mRNA X06367.1 - - 158 76

View Full Product Details