Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBQLN1
Entrez Gene ID
100302096
Gene Name
ubiquilin 1
Gene Aliases
-
Chromosome Location
Chr.2: 37901747 - 37954676 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161886.1 NP_001155358.1 2 - 3 332 79
XM_012110487.3 XP_011965877.1 2 - 3 570 79
GenBank mRNA FJ958367.1 - 2 - 3 332 79

View Full Product Details