Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD3E
Entrez Gene ID
443437
Gene Name
CD3e molecule
Gene Aliases
-
Chromosome Location
Chr.15: 31827379 - 31837998 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009418.2 NP_001009418.1 4 - 5 216 89
GenBank mRNA S53077.1 - 4 - 5 217 89
Z12969.1 - - 217 89

View Full Product Details