Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD5
Entrez Gene ID
100125355
Gene Name
abhydrolase domain containing 5, lysophosphatidic acid acyltransferase
Gene Aliases
-
Chromosome Location
Chr.19: 16584321 - 16621969 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104933.1 NP_001098403.1 2 - 3 190 108
GenBank mRNA EF660331.1 - 2 - 3 190 108

View Full Product Details