Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AHSG
Entrez Gene ID
443392
Gene Name
alpha 2-HS glycoprotein
Gene Aliases
-
Chromosome Location
Chr.1: 219108805 - 219115945 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009802.1 NP_001009802.1 5 - 6 724 83
GenBank mRNA X16578.1 - - 724 83

View Full Product Details