Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ST3GAL4
Entrez Gene ID
100127212
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 4
Gene Aliases
-
Chromosome Location
Chr.21: 30715952 - 30747543 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112818.1 NP_001106289.1 7 - 8 643 71
XM_012101523.3 XP_011956913.2 8 - 9 771 71
XM_012101525.3 XP_011956915.1 8 - 9 782 71
GenBank mRNA AM888395.1 - 7 - 8 643 71

View Full Product Details