Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS4
Entrez Gene ID
100381246
Gene Name
regulator of G protein signaling 4
Gene Aliases
-
Chromosome Location
Chr.1: 121932030 - 121936791 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174111.1 NP_001167582.1 2 - 3 147 83
GenBank mRNA FJ715737.1 - 2 - 3 147 83

View Full Product Details