Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ID3
Entrez Gene ID
100415775
Gene Name
inhibitor of DNA binding 3, HLH protein
Gene Aliases
-
Chromosome Location
Chr.2: 258829839 - 258831546 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177678.1 NP_001171149.1 1 - 2 510 68
GenBank mRNA GU437213.1 - 1 - 2 510 68

View Full Product Details