Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD40LG
Entrez Gene ID
767628
Gene Name
CD40 ligand
Gene Aliases
CD154
Chromosome Location
Chr.X: 107150981 - 107162741 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075101.1 NP_001068569.1 2 - 3 283 106
GenBank mRNA DQ054533.1 - 2 - 3 283 106

View Full Product Details