Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAG3
Entrez Gene ID
443375
Gene Name
pregnancy-associated glycoprotein 3
Gene Aliases
-
Chromosome Location
Chr.21: 39031713 - 39693216 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009792.2 NP_001009792.1 3 - 4 327 129
XM_012101545.3 XP_011956935.1 3 - 4 364 129
GenBank mRNA U94789.1 - 3 - 4 327 129

View Full Product Details