Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEAK1
Entrez Gene ID
101104347
Gene Name
pseudopodium enriched atypical kinase 1
Gene Aliases
-
Chromosome Location
Chr.18: 30208568 - 30539381 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957162.1 XP_027812963.1 7 - 8 4658 74

View Full Product Details