Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUFY4
Entrez Gene ID
101108551
Gene Name
RUN and FYVE domain containing 4
Gene Aliases
-
Chromosome Location
Chr.2: 234976024 - 234997791 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965166.1 XP_027820967.1 3 - 4 380 60

View Full Product Details