Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM72A
Entrez Gene ID
101116244
Gene Name
family with sequence similarity 72 member A
Gene Aliases
-
Chromosome Location
Chr.12: 3588334 - 3601421 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013564.4 XP_004013613.1 2 - 3 1192 82

View Full Product Details