Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NACC1
Entrez Gene ID
101121422
Gene Name
nucleus accumbens associated 1
Gene Aliases
-
Chromosome Location
Chr.5: 10832382 - 10850978 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969530.1 XP_027825331.1 2 - 3 1154 97

View Full Product Details