Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCEE
Entrez Gene ID
101117702
Gene Name
methylmalonyl-CoA epimerase
Gene Aliases
-
Chromosome Location
Chr.18: 25250673 - 25259073 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017800.4 XP_004017849.2 1 - 2 107 69

View Full Product Details