Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM9B
Entrez Gene ID
101106198
Gene Name
TMEM9 domain family member B
Gene Aliases
-
Chromosome Location
Chr.15: 46779542 - 46797762 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015101041.2 XP_014956527.2 2 - 3 302 110

View Full Product Details