Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXB13
Entrez Gene ID
101114626
Gene Name
homeobox B13
Gene Aliases
-
Chromosome Location
Chr.11: 25533313 - 25535076 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012802.3 XP_004012851.2 1 - 2 606 88

View Full Product Details