Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB38
Entrez Gene ID
101122822
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.21: 8468744 - 8534145 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019394.3 XP_004019443.1 2 - 3 527 61

View Full Product Details