Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NR2C2AP
Entrez Gene ID
101106004
Gene Name
nuclear receptor 2C2 associated protein
Gene Aliases
-
Chromosome Location
Chr.5: 4108908 - 4111184 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008406.4 XP_004008455.1 3 - 4 616 90

View Full Product Details