Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TKDP5
Entrez Gene ID
443180
Gene Name
uncharacterized TKDP5
Gene Aliases
-
Chromosome Location
Chr.13: 77227367 - 77282094 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976400.1 XP_027832201.1 31 - 32 2638 87
GenBank mRNA AF529269.1 - 16 - 17 1312 87

View Full Product Details