Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FKBP7
Entrez Gene ID
101123607
Gene Name
FKBP prolyl isomerase 7
Gene Aliases
-
Chromosome Location
Chr.2: 142272071 - 142279753 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004544.4 XP_004004593.2 2 - 3 470 95

View Full Product Details