Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C6H4orf17
Entrez Gene ID
101111454
Gene Name
chromosome 6 C4orf17 homolog
Gene Aliases
-
Chromosome Location
Chr.6: 28810350 - 28831341 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970859.1 XP_027826660.1 1 - 2 679 101

View Full Product Details