Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYLC1
Entrez Gene ID
101121662
Gene Name
cylicin 1
Gene Aliases
-
Chromosome Location
Chr.X: 81406592 - 81452795 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022224.3 XP_004022273.2 5 - 6 2150 89
XM_012106994.2 XP_011962384.2 4 - 5 2109 89

View Full Product Details