Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROPN1
Entrez Gene ID
101116109
Gene Name
rhophilin associated tail protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 206439277 - 206469494 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002985.4 XP_004003034.1 2 - 3 309 80
XM_012095901.3 XP_011951291.1 2 - 3 218 80

View Full Product Details