Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNB1IP1
Entrez Gene ID
101115966
Gene Name
cyclin B1 interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 43526701 - 43534440 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012181091.2 XP_012036481.1 2 - 3 474 72

View Full Product Details