Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF75D
Entrez Gene ID
101106826
Gene Name
zinc finger protein 75D
Gene Aliases
-
Chromosome Location
Chr.X: 108381781 - 108409832 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012107284.3 XP_011962674.2 4 - 5 1075 96
XM_012107287.3 XP_011962677.2 4 - 5 1058 96
XM_012107288.3 XP_011962678.2 4 - 5 985 96
XM_012107289.3 XP_011962679.2 4 - 5 985 96
XM_012107291.3 XP_011962681.2 4 - 5 985 96
XM_027963174.1 XP_027818975.1 4 - 5 985 96
XM_027963175.1 XP_027818976.1 4 - 5 985 96
XM_027963176.1 XP_027818977.1 4 - 5 985 96
XM_027963177.1 XP_027818978.1 3 - 4 558 96
XM_027963178.1 XP_027818979.1 3 - 4 627 96

View Full Product Details