Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSMEM1
Entrez Gene ID
101116225
Gene Name
serine rich single-pass membrane protein 1
Gene Aliases
C4H7orf45
Chromosome Location
Chr.4: 102793324 - 102800745 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008062.3 XP_004008111.2 1 - 2 181 109

View Full Product Details