Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCWPW2
Entrez Gene ID
101109307
Gene Name
zinc finger CW-type and PWWP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.19: 3000360 - 3153814 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958145.1 XP_027813946.1 7 - 8 1778 103

View Full Product Details