Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUFU
Entrez Gene ID
101114993
Gene Name
SUFU negative regulator of hedgehog signaling
Gene Aliases
-
Chromosome Location
Chr.22: 25733050 - 25843036 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020324.4 XP_004020373.4 3 - 4 607 78
XM_015103389.2 XP_014958875.2 3 - 4 608 78
XM_015103390.2 XP_014958876.2 3 - 4 608 78

View Full Product Details