Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMB5
Entrez Gene ID
101123468
Gene Name
proteasome 20S subunit beta 5
Gene Aliases
-
Chromosome Location
Chr.7: 23140494 - 23145935 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010336.4 XP_004010385.1 1 - 2 432 87

View Full Product Details