Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFPM2
Entrez Gene ID
101121179
Gene Name
zinc finger protein, FOG family member 2
Gene Aliases
-
Chromosome Location
Chr.9: 79651978 - 80174933 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973106.1 XP_027828907.1 3 - 4 328 108

View Full Product Details