Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC85A
Entrez Gene ID
101120398
Gene Name
coiled-coil domain containing 85A
Gene Aliases
-
Chromosome Location
Chr.3: 72567449 - 72787512 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005934.4 XP_004005983.3 3 - 4 1387 72
XM_027966814.1 XP_027822615.1 3 - 4 1388 72

View Full Product Details