Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LIPT2
Entrez Gene ID
101105357
Gene Name
lipoyl(octanoyl) transferase 2
Gene Aliases
-
Chromosome Location
Chr.15: 56940832 - 56942492 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979043.1 XP_027834844.1 1 - 2 449 56

View Full Product Details