Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AZU1
Entrez Gene ID
101110329
Gene Name
azurocidin 1
Gene Aliases
-
Chromosome Location
Chr.5: 45495478 - 45502402 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009470.4 XP_004009519.2 5 - 6 727 111

View Full Product Details